accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CONSOOOONNONOH3C

CAS RN 344930-95-6

IUPAC Name: 6-methoxy-2-[({4-oxo-9-[2-(piperidin-1-yl)ethoxy]-4H-pyrido[1,2-a]pyrimidin-2-yl}oxy)methyl]-4-(propan-2-yl)-2,3-dihydro-1λ⁶,2-benzothiazole-1,1,3-trione
Synonyms: 4-isopropyl-6-methoxy-2-[({4-oxo-9-[2-(piperidin-1-yl)ethoxy]pyrido[1,2-a]pyrimidin-2-yl}oxy)methyl]-1λ⁶,2-benzothiazole-1,1,3-trione
Molecular Weight (g/mol): 556.63
Molecular Formula: C27H32N4O7S
InChi Key: DRZXDZYWZSKFDL-UHFFFAOYSA-N
SMILES: COC1=CC2=C(C(=O)N(COC3=CC(=O)N4C=CC=C(OCCN5CCCCC5)C4=N3)S2(=O)=O)C(=C1)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results