Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketotifen fumarate, R&D Systems™
SDP

Catalog No. 0784100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-841-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 07-841-00 Supplier R&D Systems Supplier No. 0784/100

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. H1 antagonist

An H1 receptor antagonist.

Chemical Identifiers

CAS 34580-14-8
Molecular Formula C23H23NO5S
Molecular Weight (g/mol) 425.499
InChI Key YNQQEYBLVYAWNX-WLHGVMLRSA-N
Synonym ketotifen fumarate, zaditen, ketotifen hydrogen fumarate, alaway, unii-hbd503woro, hc 20,511 fumarate, ketotifen fumarate usan:jan, ketotifen fumarate salt, hbd503woro
PubChem CID 5282408
IUPAC Name (E)-but-2-enedioic acid;10-(1-methylpiperidin-4-ylidene)-5H-benzo[1,2]cyclohepta[3,4-b]thiophen-4-one
SMILES CN1CCC(=C2C3=C(C(=O)CC4=CC=CC=C42)SC=C3)CC1.C(=CC(=O)O)C(=O)O

Specifications

Quantity 100 mg
Formula Weight 425.5
Percent Purity >99%
Product Line Tocris
Chemical Name or Material Ketotifen fumarate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.