accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34823-86-4

NHNO

CAS RN 34823-86-4

IUPAC Name: 5-phenyl-7-oxa-3,4-diazatricyclo[6.4.0.0²,⁶]dodeca-1(12),2(6),4,8,10-pentaene
Synonyms: 5-phenyl-7-oxa-3,4-diazatricyclo[6.4.0.0²,⁶]dodeca-1(12),2(6),4,8,10-pentaene
Molecular Weight (g/mol): 234.26
Molecular Formula: C15H10N2O
InChi Key: DZQLVVLATXPWBK-UHFFFAOYSA-N
SMILES: N1N=C(C2=C1C1=CC=CC=C1O2)C1=CC=CC=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results