accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 368866-21-1

CH3CH3CH3OONOHOHNOO

CAS RN 368866-21-1

IUPAC Name: 1-[(tert-butoxy)carbonyl]-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)piperidine-3-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}piperidine-3-carboxylic acid
Molecular Weight (g/mol): 466.53
Molecular Formula: C26H30N2O6
InChi Key: VXJHDZRKJBBFKW-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCC(C1)(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results