Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-FMOC-Amino-(3-N-BOC-piperidinyl)carboxylic acid, 95%, Thermo Scientific™

Catalog No. AC360515000
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368866-21-1
Molecular Formula C26H30N2O6
Molecular Weight (g/mol) 466.53
MDL Number MFCD02683125
InChI Key VXJHDZRKJBBFKW-UHFFFAOYNA-N
Synonym 1-tert-butoxy carbonyl-3-9h-fluoren-9-ylmethoxy carbonyl amino piperidine-3-carboxylic acid, 3-9h-fluoren-9-ylmethoxycarbonylamino-piperidine-1,3-dicarboxylic acid 1-tert-butyl ester, 1-tert-butoxycarbonyl-3-9h-fluoren-9-ylmethoxy carbonyl amino piperidine-3-carboxylic acid, vxjhdzrkjbbfkw-uhfffaoysa-n, 1-boc-3-fmoc-amino-3-piperidinecarboxylic acid, 3-n-fmoc-amino-n-bocpiperidine-3-carboxylic acid, n-fmoc-amino-3-n-boc-piperidinyl carboxylic acid, 3-n-fmoc-amino-n-boc-3-piperidinyl carboxylic acid, 1-tert-butoxy carbonyl-3-9h-fluoren-9-yl methoxy carbonyl amino piperidine-3-carboxylic acid
PubChem CID 2734408

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 500 mg
Formula Weight 466.53
Percent Purity ≥94%
Physical Form Crystalline Powder
Chemical Name or Material N-FMOC-Amino-(3-N-BOC-piperidinyl)carboxylic acid, 95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.