CAS RN 37739-05-2
IUPAC Name:
(2R,3R,4S,5R)-2-[2-chloro-6-(cyclopentylamino)-9H-purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms:
ccpa (biochemistry)
Molecular Weight (g/mol):
369.81
Molecular Formula:
C15H20ClN5O4
InChi Key:
XSMYYYQVWPZWIZ-IDTAVKCVSA-N
SMILES:
OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(NC3CCCC3)N=C(Cl)N=C12
1
–
1
of
1
results
1
R&D Systems™ 2-Chloro-N6-cyclopentyladenosine
CAS: 37739-05-2 Molecular Formula: C15H20ClN5O4