accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38598-34-4

OOAlOOOH

CAS RN 38598-34-4

IUPAC Name: [(4-cyclohexylbutanoyl)oxy](hydroxy)alumanyl 4-cyclohexylbutanoate
Synonyms: [(4-cyclohexylbutanoyl)oxy](hydroxy)alumanyl 4-cyclohexylbutanoate
Molecular Weight (g/mol): 382.48
Molecular Formula: C20H35AlO5
InChi Key: JUGYFOMQOWSWDP-UHFFFAOYSA-K
SMILES: O[Al](OC(=O)CCCC1CCCCC1)OC(=O)CCCC1CCCCC1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results