accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH3Cl

CAS RN 41965-95-1

IUPAC Name: (3-chloro-4-methoxyphenyl)methanaminium
Synonyms: (3-chloro-4-methoxyphenyl)methanaminium
Molecular Weight (g/mol): 172.63
Molecular Formula: C8H11ClNO
InChi Key: OCNMSDZALRAYEX-UHFFFAOYSA-O
SMILES: COC1=CC=C(C[NH3+])C=C1Cl
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)