accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 422513-13-1

IUPAC Name: N-{2-oxo-3-[phenyl({4-[(piperidin-1-yl)methyl]phenyl}amino)methylidene]-2,3-dihydro-1H-indol-5-yl}ethane-1-sulfonamide
Synonyms: hesperadin
Molecular Weight (g/mol): 516.66
Molecular Formula: C29H32N4O3S
InChi Key: GLDSKRNGVVYJAB-UHFFFAOYSA-N
SMILES: CCS(=O)(=O)NC1=CC2=C(NC(=O)C2=C(NC2=CC=C(CN3CCCCC3)C=C2)C2=CC=CC=C2)C=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results