accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 43077-30-1

IUPAC Name: {[5-methyl-2-(propan-2-yl)cyclohexyl](phenyl)phosphoroso}benzene
Synonyms: [2-isopropyl-5-methylcyclohexyl(phenyl)phosphoroso]benzene
Molecular Weight (g/mol): 340.45
Molecular Formula: C22H29OP
InChi Key: BPCNGVCAHAIZEE-UHFFFAOYNA-N
SMILES: CC(C)C1CCC(C)CC1P(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results