accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOOSOONCH3CH3

CAS RN 433695-36-4

IUPAC Name: N,N,2-trimethyl-5-nitrobenzene-1-sulfonamide
Synonyms: N,N,2-trimethyl-5-nitrobenzenesulfonamide
Molecular Weight (g/mol): 244.27
Molecular Formula: C9H12N2O4S
InChi Key: IFIUFCJFLGCQPH-UHFFFAOYSA-N
SMILES: CN(C)S(=O)(=O)C1=C(C)C=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results