CAS RN 467-63-0
CAS RN 467-63-0
IUPAC Name:
tris[4-(dimethylamino)phenyl]methanol
Synonyms:
tris[4-(dimethylamino)phenyl]methanol
Molecular Weight (g/mol):
389.54
Molecular Formula:
C25H31N3O
InChi Key:
QFVDKARCPMTZCS-UHFFFAOYSA-N
SMILES:
CN(C)C1=CC=C(C=C1)C(O)(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C