Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crystal Violet Gram Solution MilliporeSigma™

Catalog No. M65092A85 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
Catalog No. Quantity
M65092A85 4 L
1 options

Catalog No. M65092A85

Supplier: MilliporeSigma 65092A85

Crystal Violet; Gram Solution; HARLECO; Stains Gram-positive bacteria purple

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 467-63-0

Specifications

Packaging Poly Bottle
Quantity 4 L
Chemical Name or Material Crystal Violet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.