accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNOONHNCH3CH3CH3OH3COOH2NH3CH3COHOCH3HCl(E/Z)(E/Z)(E/Z)

CAS RN 467214-21-7

IUPAC Name: 19-{[2-(dimethylamino)ethyl]amino}-13-hydroxy-8,14-dimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate hydrochloride
Synonyms: 19-{[2-(dimethylamino)ethyl]amino}-13-hydroxy-8,14-dimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate hydrochloride
Molecular Weight (g/mol): 653.21
Molecular Formula: C32H49ClN4O8
InChi Key: DFSYBWLNYPEFJK-UHFFFAOYNA-N
SMILES: Cl.COC1CC(C)CC2=C(NCCN(C)C)C(=O)C=C(NC(=O)C(C)=CC=CC(OC)C(OC(N)=O)C(C)=CC(C)C1O)C2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results