accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 468740-43-4

IUPAC Name: 4-{[2-(3-chlorophenyl)-2-hydroxyethyl]amino}-3-[4-methyl-6-(morpholin-4-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-ylidene]-2,3-dihydropyridin-2-one
Synonyms: 4-{[2-(3-chlorophenyl)-2-hydroxyethyl]amino}-3-[4-methyl-6-(morpholin-4-yl)-1,3-dihydro-1,3-benzodiazol-2-ylidene]pyridin-2-one
Molecular Weight (g/mol): 479.97
Molecular Formula: C25H26ClN5O3
InChi Key: UGQMURPIMYALPH-UHFFFAOYNA-N
SMILES: CC1=CC(=CC2=C1NC(N2)=C1C(=O)N=CC=C1NCC(O)C1=CC(Cl)=CC=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results