CAS RN 487-79-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscience™ Kainic acid (synthetic)
Kainate agonist; excitant and neurotoxin
2
MilliporeSigma™ Calbiochem™ Kainic Acid
An excitatory amino acid receptor agonist selective for the kainate receptor subtype (Ki = 21nM for 3H-kainate binding in rat striatum).
3
Tocris Bioscience™ Kainic acid
Kainate agonist; excitant and neurotoxin