accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNOOHOSF(E/Z)

CAS RN 494191-73-0

IUPAC Name: 4-({2-[(2-fluorophenyl)amino]-4-oxo-4,5-dihydro-1,3-thiazol-5-ylidene}methyl)benzoic acid
Synonyms: 4-({2-[(2-fluorophenyl)amino]-4-oxo-1,3-thiazol-5-ylidene}methyl)benzoic acid
Molecular Weight (g/mol): 342.34
Molecular Formula: C17H11FN2O3S
InChi Key: SYCKPHBALHXMIR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C2SC(NC3=CC=CC=C3F)=NC2=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)