CAS RN 495076-64-7
IUPAC Name:
4-(benzyloxy)-N-{[1-(dimethylamino)cyclopentyl]methyl}-3,5-dimethoxybenzamide hydrochloride
Synonyms:
4-(benzyloxy)-N-{[1-(dimethylamino)cyclopentyl]methyl}-3,5-dimethoxybenzamide hydrochloride
Molecular Weight (g/mol):
448.99
Molecular Formula:
C24H33ClN2O4
InChi Key:
NIPQJILJYQVZJR-UHFFFAOYSA-N
SMILES:
Cl.COC1=CC(=CC(OC)=C1OCC1=CC=CC=C1)C(=O)NCC1(CCCC1)N(C)C
1
–
1
of
1
results
1
R&D Systems™ Org 25543 hydrochloride
CAS: 495076-64-7 Molecular Formula: C24H33ClN2O4