Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Niclosamide
GREENER_CHOICESDP

Catalog No. 407950 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-795-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 40-795-0 Supplier R&D Systems™ Supplier No. 4079/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. STAT3 inhibitor; also inhibits mTORC1 signaling

Inhibitor of the STAT3 signaling pathway; inhibits the activation, nuclear translocation and transactivation of STAT3. Displays selectivity for STAT3 over STAT1, STAT5, JAK1, JAK2 and Src kinases. Inhibits the transcription of STAT3 target genes and induces cell growth inhibition, apoptosis and cell cycle arrest of cancer cells with constitutively active STAT3. Also reversibly inhibits mTORC1 signaling and stimulates autophagy in vitro; displays antineoplastic effects in acute myelogenous leukemia (AML) stem cells.

Chemical Identifiers

CAS 50-65-7
Molecular Formula C13H8Cl2N2O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 325.99
Purity >99%
Molecular Weight (g/mol) 327.12
Chemical Name or Material Niclosamide
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.