CAS RN 50-76-0
Actinomycin D (1 mg/ml, DMSO)
CAS: 50-76-0 Molecular Formula: C62H86N12O16 Molecular Weight (g/mol): 1255.438 MDL Number: MFCD00005033 InChI Key: RJURFGZVJUQBHK-HUZQGMAJSA-N Synonym: Dactinomycin; Actinomycin C1 PubChem CID: 131954673 IUPAC Name: 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4, SMILES: CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C
Actinomycin D, >99%, MP Biomedicals™
CAS: 50-76-0 Molecular Formula: C62H86N12O16 Molecular Weight (g/mol): 1255.438 InChI Key: RJURFGZVJUQBHK-HUZQGMAJSA-N Synonym: Dactinomycin,Actinomycin-[thr-val-pro-sar-meval] PubChem CID: 131954673 IUPAC Name: 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4, SMILES: CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C
Actinomycin D
CAS: 50-76-0 Molecular Formula: C62H86N12O16 Molecular Weight (g/mol): 1255.438 MDL Number: MFCD00005033 InChI Key: RJURFGZVJUQBHK-HUZQGMAJSA-N Synonym: Dactinomycin; Actinomycin C1 PubChem CID: 131954673 IUPAC Name: 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4, SMILES: CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C
Thermo Scientific Chemicals Actinomycin D, 93%
CAS: 50-76-0 Molecular Formula: C62H86N12O16 Molecular Weight (g/mol): 1255.438 MDL Number: MFCD00005033 InChI Key: RJURFGZVJUQBHK-HUZQGMAJSA-N Synonym: Dactinomycin PubChem CID: 131954673 IUPAC Name: 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4, SMILES: CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C
Actinomycin D Crystalline MP Biomedicals
CAS: 50-76-0 Molecular Formula: C62H86N12O16 Molecular Weight (g/mol): 1255.438 MDL Number: MFCD00005033 InChI Key: RJURFGZVJUQBHK-HUZQGMAJSA-N Synonym: Dactinomycin,actinomycin-[thr-val-pro-sar-meval] PubChem CID: 131954673 IUPAC Name: 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4, SMILES: CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C
R&D Systems™ Actinomycin D
CAS: 50-76-0 Molecular Formula: C62H86N12O16
Novus Biologicals™ Actinomycin D Inhibitor
Low molecular weight, organic compound that may help regulate biological process.