Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Actinomycin D
GREENER_CHOICESDP

Catalog No. 122910 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-291-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-291-0 Supplier R&D Systems™ Supplier No. 1229/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibits RNA polymerase

Actinomycin D is an anti-neoplastic antibiotic. Inhibits RNA polymerase and is a potent inducer of apoptosis.

Chemical Identifiers

CAS 50-76-0
Molecular Formula C62H86N12O16

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 1255.43
Product Type Actinomycin D
Purity 95%
Molecular Weight (g/mol) 1255.438
Chemical Name or Material Actinomycin D
Recommended Storage Store at -20°C, Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.