accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONHOHNOCH3OCH3Cl

CAS RN 501925-31-1

IUPAC Name: 1-(5-chloro-2,4-dimethoxyphenyl)-3-(5-methyl-1,2-oxazol-3-yl)urea
Synonyms: 1-(5-chloro-2,4-dimethoxyphenyl)-3-(5-methyl-1,2-oxazol-3-yl)urea
Molecular Weight (g/mol): 311.72
Molecular Formula: C13H14ClN3O4
InChi Key: CEIIEALEIHQDBX-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(Cl)C=C1NC(=O)NC1=NOC(C)=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results