Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 4-Aminopyridine
GREENER_CHOICESDP

Catalog No. 0940100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-401-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 09-401-00 Supplier R&D Systems™ Supplier No. 0940/100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Non-selective KV channel blocker

Non-selective voltage-dependent K+-channel blocker (IC50 values are 170 and 230 μM at KV1.1 and KV1.2 respectively). Caged 4-AP (Cat. No. 3557) is also available.

Chemical Identifiers

CAS 504-24-5
Molecular Formula C5H6N2

Specifications

Quantity 100 mg
Product Line Tocris
Formula Weight 94.12
Purity >99%
Molecular Weight (g/mol) 94.117
Chemical Name or Material 4-Aminopyridine
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.