accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51639-48-6

H3CONNH2

CAS RN 51639-48-6

IUPAC Name: 4-(4-acetylphenyl)piperazin-1-ium
Synonyms: 4-(4-acetylphenyl)piperazin-1-ium
Molecular Weight (g/mol): 205.28
Molecular Formula: C12H17N2O
InChi Key: KPXVKKBJROCIJB-UHFFFAOYSA-O
SMILES: CC(=O)C1=CC=C(C=C1)N1CC[NH2+]CC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results