CAS RN 5204-74-0
CAS RN 5204-74-0
IUPAC Name:
acetic acid; triethylamine
Synonyms:
triethylammonium acetate
Molecular Weight (g/mol):
161.25
Molecular Formula:
C8H19NO2
InChi Key:
AVBGNFCMKJOFIN-UHFFFAOYSA-N
SMILES:
CC(O)=O.CCN(CC)CC
1
–
4
of
4
results
1
Honeywell Fluka™ Triethylammonium acetate buffer, BioUltra, volatile buffer, ∼1.0 M in H2O, Honeywell Fluka™
BioUltra, volatile buffer, ∼1.0 M in H2O
2
LiChropur™ Acetic Acid - Triethylamine Solution 1:1, MilliporeSigma™ Supelco™
CAS: 67-56-1 Molecular Formula: C8H19NO2 Molecular Weight (g/mol): 161.25 MDL Number: MFCD00151725 InChI Key: AVBGNFCMKJOFIN-UHFFFAOYSA-N Synonym: Triethylamine : Acetic acid 1:1 solution; Triethylammonium acetate solution IUPAC Name: acetic acid; triethylamine SMILES: CC(O)=O.CCN(CC)CC
3
MilliporeSigma™ Triethylammonium Acetate, 1 M Solution, Calbiochem™,

Ready-to-use buffer solution used for the purification of chemically synthesized oligonucleotides by HPLC