CAS RN 524-12-9
CAS RN 524-12-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ IKK Inhibitor II, Wedelolactone
The IKK Inhibitor II, Wedelolactone, also referenced under CAS 65646-68-6, controls the biological activity of IKK. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.