accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)OH(S)(S)(S)OOHO(E)(Z)

CAS RN 56985-40-1

IUPAC Name: (5Z)-7-[(1S,4S,6S)-6-[(1E,3S)-3-hydroxyoct-1-en-1-yl]-2-oxabicyclo[2.2.1]heptan-5-yl]hept-5-enoic acid
Synonyms: (5Z)-7-[(1S,4S,6S)-6-[(1E,3S)-3-hydroxyoct-1-en-1-yl]-2-oxabicyclo[2.2.1]heptan-5-yl]hept-5-enoic acid
Molecular Weight (g/mol): 350.5
Molecular Formula: C21H34O4
InChi Key: LQANGKSBLPMBTJ-WTPHSRJXSA-N
SMILES: CCCCC[C@H](O)\C=C\[C@@H]1[C@@H]2C[C@H](CO2)C1C\C=C/CCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)