accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONHNOHNCl

CAS RN 57653-26-6

IUPAC Name: 1-(3-chlorophenyl)-3-(1-methyl-4-oxo-4,5-dihydro-1H-imidazol-2-yl)urea
Synonyms: fenobam 非诺班
Molecular Weight (g/mol): 266.69
Molecular Formula: C11H11ClN4O2
InChi Key: DWPQODZAOSWNHB-UHFFFAOYSA-N
SMILES: CN1CC(=O)N=C1NC(=O)NC1=CC=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)