Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Clozapine
GREENER_CHOICESDP

Catalog No. 0444500 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-445-00 500 mg
04-445-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 04-445-00 Supplier R&D Systems™ Supplier No. 0444/500

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Dopamine antagonist with some D4 selectivity; also 5-HT2A/2C antagonist

Clozapine is an atypical antipsychotic drug, with a much lower tendency to cause extrapyramidal side effects than conventional neuroleptics. Displays a broad range of pharmacological actions; the antipsychotic effects are thought to be mediated principally by 5-HT2A/2C and dopamine receptor blockade (Ki values are 21, 170, 170, 230 and 330 nM for D4, D3, D1, D2 and D5 receptors respectively).

Chemical Identifiers

CAS 5786-21-0
Molecular Formula C18H19ClN4

Specifications

Quantity 500 mg
Product Line Tocris
Formula Weight 326.83
Purity >99%
Molecular Weight (g/mol) 326.828
Chemical Name or Material Clozapine
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.