accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHNONNNNH

CAS RN 612847-09-3

IUPAC Name: 1-{1-[(4-{6-phenyl-1H-imidazo[4,5-g]quinoxalin-7-yl}phenyl)methyl]piperidin-4-yl}-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 1-{1-[(4-{7-phenyl-3H-imidazo[4,5-g]quinoxalin-6-yl}phenyl)methyl]piperidin-4-yl}-3H-1,3-benzodiazol-2-one
Molecular Weight (g/mol): 551.65
Molecular Formula: C34H29N7O
InChi Key: BIWGYFZAEWGBAL-UHFFFAOYSA-N
SMILES: O=C1NC2=CC=CC=C2N1C1CCN(CC2=CC=C(C=C2)C2=C(N=C3C=C4N=CNC4=CC3=N2)C2=CC=CC=C2)CC1
Form 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results