accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONHBrBrOH(E/Z)

CAS RN 62004-35-7

IUPAC Name: 2-cyano-N-(2,5-dibromophenyl)-3-hydroxybut-2-enamide
Synonyms: 2-cyano-N-(2,5-dibromophenyl)-3-hydroxybut-2-enamide
Molecular Weight (g/mol): 360.01
Molecular Formula: C11H8Br2N2O2
InChi Key: UVSVTDVJQAJIFG-UHFFFAOYSA-N
SMILES: CC(O)=C(C#N)C(=O)NC1=C(Br)C=CC(Br)=C1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results