accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHOHOHNO

CAS RN 622864-54-4

IUPAC Name: 9-hydroxy-4-phenyl-1H,2H,3H,6H-pyrrolo[3,4-c]carbazole-1,3-dione
Synonyms: 9-hydroxy-4-phenyl-2H,6H-pyrrolo[3,4-c]carbazole-1,3-dione
Molecular Weight (g/mol): 328.33
Molecular Formula: C20H12N2O3
InChi Key: IAUZTOZLTFSMIE-UHFFFAOYSA-N
SMILES: OC1=CC2=C(NC3=C2C2=C(C(=O)NC2=O)C(=C3)C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results