accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6291-85-6

IUPAC Name: 3-ethoxypropan-1-amine
Synonyms: 1-propanamine, 3-ethoxy-
Molecular Weight (g/mol): 103.17
Molecular Formula: C5H13NO
InChi Key: SOYBEXQHNURCGE-UHFFFAOYSA-N
SMILES: CCOCCCN
Boiling Point 
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)