CAS RN 630117-19-0
IUPAC Name:
N-{[4-(2-cyanophenyl)piperazin-1-yl]methyl}-3-methylbenzamide; but-2-enedioic acid
Synonyms:
N-{[4-(2-cyanophenyl)piperazin-1-yl]methyl}-3-methylbenzamide; butenedioic acid
Molecular Weight (g/mol):
450.5
Molecular Formula:
C24H26N4O5
InChi Key:
NAEUGRPISCANHO-UHFFFAOYSA-N
SMILES:
OC(=O)C=CC(O)=O.CC1=CC=CC(=C1)C(=O)NCN1CCN(CC1)C1=CC=CC=C1C#N