CAS RN 630420-16-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Asunaprevir
Potent hepatitis C virus (HCV) NS3 protease inihibitor