accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64267-12-5

CH3OOOO

CAS RN 64267-12-5

IUPAC Name: 3-benzoyl-7-methoxy-2H-chromen-2-one
Synonyms: 3-benzoyl-7-methoxychromen-2-one
Molecular Weight (g/mol): 280.28
Molecular Formula: C17H12O4
InChi Key: HYORIVUCOQKMOC-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=C(C(=O)C3=CC=CC=C3)C(=O)OC2=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results