Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-benzoyl-7-methoxycoumarin, Thermo Scientific™

Catalog No. AC402050050
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64267-12-5
Molecular Formula C17H12O4
Molecular Weight (g/mol) 280.279
MDL Number MFCD00050046
InChI Key HYORIVUCOQKMOC-UHFFFAOYSA-N
Synonym 3-benzoyl-7-methoxycoumarin, 3-benzoyl-7-methoxy-2h-chromen-2-one, 3-benzoyl-7-methoxy coumarin, 7-methoxy-3-phenylcarbonyl-2h-chromen-2-one, 3-benzoyl-7-methoxy-chromen-2-one, 3-benzoyl-7-methoxy-1-benzopyran-2-one, pubchem13308, 7-methoxy-3-benzoylcoumarin, 7-methoxy-3-phenylcarbonyl chromen-2-one, 3-benzoyl-7-methoxy-2h-1-benzopyran-2-one
PubChem CID 342533
IUPAC Name 3-benzoyl-7-methoxychromen-2-one
SMILES COC1=CC2=C(C=C1)C=C(C(=O)O2)C(=O)C3=CC=CC=C3

Specifications

Melting Point 150°C to 153°C
Color Green to Yellow
Infrared Spectrum Authentic
Packaging Glass Bottle
Quantity 5 g
Formula Weight 280.28
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzoyl-7-methoxycoumarin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.