accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNNSONNFFF

CAS RN 659730-32-2

IUPAC Name: N-[4-({6-[4-(trifluoromethyl)phenyl]pyrimidin-4-yl}oxy)-1,3-benzothiazol-2-yl]acetamide
Synonyms: N-[4-({6-[4-(trifluoromethyl)phenyl]pyrimidin-4-yl}oxy)-1,3-benzothiazol-2-yl]acetamide
Molecular Weight (g/mol): 430.41
Molecular Formula: C20H13F3N4O2S
InChi Key: YUTIXVXZQIQWGY-UHFFFAOYSA-N
SMILES: CC(=O)NC1=NC2=C(OC3=NC=NC(=C3)C3=CC=C(C=C3)C(F)(F)F)C=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)