Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Forskolin
GREENER_CHOICESDP

Catalog No. 109950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-995-0 50 mg
10-991-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-995-0 Supplier R&D Systems™ Supplier No. 1099/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Adenylyl cyclase activator

Forskolin, or coleonol, is a cell-permeable, potent, reversible and rapid activator of adenylyl cyclase, an enzyme that converts ATP to cAMP and pyrophosphate (EC50 = 0.5 μM). In combination with 5 other compounds, forskolin enables chemical reprogramming of mouse embryonic fibroblasts to pluripotent stem cells, without genetic factors: Valproic acid, sodium salt (Cat. No. 2815), RepSox (Cat. No. 3742), Tranylcypromine hydrochloride (Cat. No. 3852), CHIR 99021 (Cat. No. 4423) and 3-Deazaneplanocin A hydrochloride (Cat. No. 4703). In combination with IBMX (Cat. No. 2845), forskolin induces neural differentiation of mesenchymal stem cells by downregulating neuron restrictive silencer factor. In a rat model of Huntington's disease-like neurodegeneration, forskolin also exhibits neuroprotective effects. Hypotensive and vasodilatory agent.

Water-soluble analog, NKH 477 (Cat. No. 1603), and Forskolin synthesized to Ancillary Material Grade also available.

For more information about how Forskolin may be used, see our protocol: Transdifferentiating Fibroblasts into Neurons.

Chemical Identifiers

CAS 66575-29-9
Molecular Formula C22H34O7

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 410.51
Purity >98%
Molecular Weight (g/mol) 410.507
Chemical Name or Material (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-5-(Acetyloxy)-3-ethenyldodecahydro-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1H-naphtho[2,1-b]pyran-1-one
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.