accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67-99-2

H3CNO(R)(S)HO(S)NO(R)OHSS

CAS RN 67-99-2

IUPAC Name: (1R,7S,8S,11R)-7-hydroxy-11-(hydroxymethyl)-15-methyl-12,13-dithia-9,15-diazatetracyclo[9.2.2.0¹,⁹.0³,⁸]pentadeca-3,5-diene-10,14-dione
Synonyms: gliotoxin gliotoxins aspergillin glitoxin
Molecular Weight (g/mol): 326.39
Molecular Formula: C13H14N2O4S2
InChi Key: FIVPIPIDMRVLAY-RBJBARPLSA-N
SMILES: CN1C(=O)[C@]23CC4=CC=C[C@H](O)[C@H]4N2C(=O)[C@@]1(CO)SS3
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results