accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHONCH3CH3(Z)

CAS RN 68047-06-3

IUPAC Name: 4-[(1Z)-1-{4-[2-(dimethylamino)ethoxy]phenyl}-1-phenylbut-1-en-2-yl]phenol
Synonyms: 4'-hydroxytamoxifen
Molecular Weight (g/mol): 387.52
Molecular Formula: C26H29NO2
InChi Key: DODQJNMQWMSYGS-QPLCGJKRSA-N
SMILES: CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=C(O)C=C1
Color 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)