accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNONNOCH3SOOCH3

CAS RN 680590-49-2

IUPAC Name: N-ethyl-2-[N-(6-methoxypyridin-3-yl)2-methylbenzenesulfonamido]-N-[(pyridin-3-yl)methyl]acetamide
Synonyms: empa (drug)
Molecular Weight (g/mol): 454.55
Molecular Formula: C23H26N4O4S
InChi Key: KJPHTXTWFHVJIG-UHFFFAOYSA-N
SMILES: CCN(CC1=CN=CC=C1)C(=O)CN(C1=CN=C(OC)C=C1)S(=O)(=O)C1=CC=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)