CAS RN 68285-23-4
CAS RN 68285-23-4
IUPAC Name:
1-(2-methoxyphenyl)ethan-1-amine
Synonyms:
1-(2-methoxyphenyl)ethanamine
Molecular Weight (g/mol):
151.21
Molecular Formula:
C9H13NO
InChi Key:
VENQOHAPVLVQKV-UHFFFAOYNA-N
SMILES:
COC1=CC=CC=C1C(C)N
1
–
1
of
1
results
1
Thermo Scientific Chemicals (R)-1-(2-Methoxyphenyl)ethylamine, ChiPros™, 99%, ee 98+%
CAS: 68285-23-4 Molecular Formula: C9H13NO Molecular Weight (g/mol): 151.21 MDL Number: MFCD00671657 InChI Key: VENQOHAPVLVQKV-UHFFFAOYNA-N Synonym: r-1-2-methoxyphenyl ethanamine,r-1-2-methoxyphenyl ethylamine,1r-1-2-methoxyphenyl ethanamine,1r-1-2-methoxyphenyl ethan-1-amine,r-2-methoxy-,a-methylbenzylamine,1r-1-2-methoxyphenyl ethylamine,r-2-methoxy-alpha-methylbenzylamine,r-+-2-methoxy a-methylbenzylamine,r-1-2-methoxyphenyl ethylamine, chipros,r-2-methoxy-alpha-methylbenzylamine, chipros r , produced by basf PubChem CID: 853083 IUPAC Name: (1R)-1-(2-methoxyphenyl)ethanamine SMILES: COC1=CC=CC=C1C(C)N