Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(R)-1-(2-Methoxyphenyl)ethylamine, ChiPros™, 99%, ee 98+%
Click to view available options
Quantity:
1 g
Description
The derivatives of the compound, are used as a potential ligand in developing radioiodinated antagonists for imaging of 5-HT1A receptors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The derivatives of the compound, are used as a potential ligand in developing radioiodinated antagonists for imaging of 5-HT1A receptors.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
The derivatives of the compound, are used as a potential ligand in developing radioiodinated antagonists for imaging of 5-HT1A receptors.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Boiling Point | 118°C (17 mmHg) |
| Quantity | 1 g |
| UN Number | UN2735 |
| Sensitivity | Air Sensitive |
| Solubility Information | Soluble in dimethyl sulfoxide. |
| Formula Weight | 151.21 |
| Percent Purity | 99% |
| Assay | ee ≥98% |
| Chemical Name or Material | (R)-1-(2-Methoxyphenyl)ethylamine |
Safety and Handling
| Hazard Category | H314-H331-H335 |
| Hazard Statement | GHS H Statement H301-H311-H314-H318 Toxic if swallowed. Toxic in contact with skin. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S. |
| TSCA | No |
| Recommended Storage | Store under Argon |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction