accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHNNNNNClClOH2NHCl

CAS RN 686347-12-6

IUPAC Name: 1-[8-(2-chlorophenyl)-9-(4-chlorophenyl)-9H-purin-6-yl]-4-(ethylamino)piperidine-4-carboxamide hydrochloride
Synonyms: motavizumab
Molecular Weight (g/mol): 546.88
Molecular Formula: C25H26Cl3N7O
InChi Key: KPYUQCJBZGQHPL-UHFFFAOYSA-N
SMILES: Cl.CCNC1(CCN(CC1)C1=C2N=C(N(C2=NC=N1)C1=CC=C(Cl)C=C1)C1=CC=CC=C1Cl)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results