CAS RN 7077-05-6
CAS RN 7077-05-6
IUPAC Name:
4-(propan-2-yl)cyclohexane-1-carboxylic acid
Synonyms:
4-isopropylcyclohexane-1-carboxylic acid
Molecular Weight (g/mol):
170.25
Molecular Formula:
C10H18O2
InChi Key:
YRQKWRUZZCBSIG-UHFFFAOYSA-N
SMILES:
CC(C)C1CCC(CC1)C(O)=O
1
–
1
of
1
results
1
trans-4-Isopropylcyclohexanecarboxylic Acid 98.0+%, TCI America™

CAS: 7077-05-6 Molecular Formula: C10H18O2 Molecular Weight (g/mol): 170.25 MDL Number: MFCD04004145 InChI Key: YRQKWRUZZCBSIG-UHFFFAOYSA-N PubChem CID: 81526 IUPAC Name: 4-propan-2-ylcyclohexane-1-carboxylic acid SMILES: CC(C)C1CCC(CC1)C(=O)O