accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71125-46-7

NNNSH2N

CAS RN 71125-46-7

IUPAC Name: 5-(piperidin-1-yl)-1,3,4-thiadiazol-2-amine
Synonyms: 5-(piperidin-1-yl)-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 184.26
Molecular Formula: C7H12N4S
InChi Key: ULKCIZIQEBWQFJ-UHFFFAOYSA-N
SMILES: NC1=NN=C(S1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results