Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-piperidin-1-yl-1,3,4-thiadiazol-2-amine, Thermo Scientific™

Catalog No. MAY00307DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAY00307DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAY00307DA Supplier Thermo Scientific Chemicals Supplier No. MAY00307DA
Only null left
Add to Cart

Chemical Identifiers

CAS 71125-46-7
Molecular Formula C7H12N4S
Molecular Weight (g/mol) 184.261
InChI Key ULKCIZIQEBWQFJ-UHFFFAOYSA-N
Synonym 5-piperidin-1-yl-1,3,4-thiadiazol-2-amine, tos-bb-1081, 2-amino-5-piperidino-1,3,4-thiadiazole, 5-piperidyl-1,3,4-thiadiazole-2-ylamine, 2-amino-5-piperid-1-yl-1,3,4-thiadiazole, 1,3,4-thiadiazol-2-amine, 5-1-piperidinyl
PubChem CID 1500010
IUPAC Name 5-piperidin-1-yl-1,3,4-thiadiazol-2-amine
SMILES C1CCN(CC1)C2=NN=C(S2)N

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 184.26
Percent Purity 97%
Chemical Name or Material 5-piperidin-1-yl-1, 3, 4-thiadiazol-2-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.