CAS RN 72214-67-6
CAS RN 72214-67-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Rb/Raf-1 Disruptor 251
A cell-permeable and orally active isothiouronium compound that potently and selectively disrupts the Rb/Raf-1 interaction, while exhibiting no effect against Rb/B-Raf, Rb/E2F1, Rb/HDAC1, Raf-1/MEK1.