accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 72845-94-4

NH3HONH3HOHONOOSOOOSOOOO

CAS RN 72845-94-4

IUPAC Name: bis(2-hydroxyethan-1-aminium) 14-(2-hydroxyethyl)-13,15-dioxo-8-oxa-14-azapentacyclo[10.6.2.0²,⁷.0⁹,¹⁹.0¹⁶,²⁰]icosa-1(18),2,4,6,9,11,16,19-octaene-4,6-disulfonate
Synonyms: bis(ethanolaminium cation) 14-(2-hydroxyethyl)-13,15-dioxo-8-oxa-14-azapentacyclo[10.6.2.0²,⁷.0⁹,¹⁹.0¹⁶,²⁰]icosa-1(18),2,4,6,9,11,16,19-octaene-4,6-disulfonate
Molecular Weight (g/mol): 613.61
Molecular Formula: C24H27N3O12S2
InChi Key: QVBXNCYILVOQRY-UHFFFAOYSA-N
SMILES: [NH3+]CCO.[NH3+]CCO.OCCN1C(=O)C2=CC=C3OC4=C(C=C(C=C4C4=CC=C(C1=O)C2=C34)S([O-])(=O)=O)S([O-])(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)